Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
CAS: 542-85-8 | C3H5NS | 87.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR376860250
Price (MYR)
1,300.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,300.00
Add to cart
Chemical Identifiers
CAS6104-58-1
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular FormulaC47H48N3NaO7S2
InChI KeyCMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Dye)≥65%
CommentSuitable for protein staining
Solution Test1 mg/ml in water produces a dark blue solution
Appearance (Color)Dark blue to blue-black or dark purple or dark brown to violet brown