DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
CAS: 95-96-5 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR376851000
Price (MYR)
1,859.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,859.00
Add to cart
Chemical Identifiers
CAS3262-72-4
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular FormulaC8H15NO5
InChI KeyFHOAKXBXYSJBGX-MLHKIVSYNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-7° to -8° (20°C, 589 nm) (c=2, H2O)
Water7.5 to 8.0 % (K.F.)
Titration with NaOH>=97.5 %
Appearance (Color)White
Appearance (Form)Powder