DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
CAS: 95-96-5 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR376850250
Price (MYR)
767.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
767.00
Add to cart
Chemical Identifiers
CAS534-17-8
IUPAC Namedicaesium(1+) carbonate
Molecular FormulaCCs2O3
InChI KeyFJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total trace metal impurities=<0.01 %
Appearance (Form)Powder