3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR376030250
Price (MYR)
4,719.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,719.00
Add to cart
Chemical Identifiers
CAS7601-90-3
IUPAC Nameperchloric acid
Molecular FormulaClHO4
InChI KeyVLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILESO[Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay from Supplier's CofA0.1N