1-Methylpyrazole, 98%, Thermo Scientific Chemicals
1-Methylpyrazole, 98%, Thermo Scientific Chemicals
1-Methylpyrazole, 98%, Thermo Scientific Chemicals
1-Methylpyrazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylpyrazole, 98%, Thermo Scientific Chemicals
CAS: 930-36-9 | C4H6N2 | 82.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR373540500
Price (MYR)
1,843.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,843.00
Add to cart
Chemical Identifiers
CAS80-58-0
IUPAC Name2-bromobutanoic acid
Molecular FormulaC4H7BrO2
InChI KeyYAQLSKVCTLCIIE-UHFFFAOYNA-N
SMILESCCC(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4715-1.4755 @ 20?C
Assay (GC)≥97.5%