4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR373300250
Price (MYR)
2,637.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,637.00
Add to cart
Chemical Identifiers
CAS20980-22-7
IUPAC Name2-(piperazin-1-yl)pyrimidine
Molecular FormulaC8H12N4
InChI KeyMRBFGEHILMYPTF-UHFFFAOYSA-N
SMILESC1CN(CCN1)C1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Transition TemperatureWhite to yellow or clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.5865-1.5895 @ 20?C
Appearance (Color)Clear
FormPowder or fused solid or liquid as melt