4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals
CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR373300050
Price (MYR)
777.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
777.00
Add to cart
Chemical Identifiers
CAS79-39-0
IUPAC Name2-methylprop-2-enamide
Molecular FormulaC4H7NO
InChI KeyFQPSGWSUVKBHSU-UHFFFAOYSA-N
SMILESCC(=C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Color)White to light yellow
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point108°C to 112°C