Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
CAS: 122-78-1 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR370915000
Price (MYR)
1,255.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,255.00
Add to cart
Chemical Identifiers
CAS16473-35-1
IUPAC Name[4-(chloromethyl)phenyl]methanol
Molecular FormulaC8H9ClO
InChI KeyOGALXJIOJZXBBP-UHFFFAOYSA-N
SMILESOCC1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
GC>=96.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point57°C to 61°C