Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
CAS: 122-78-1 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR370911000
Price (MYR)
496.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Chemical Identifiers
CAS373-57-9
IUPAC Nametrifluoroborane
Molecular FormulaBF3
InChI KeyWTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILESFB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to amber
Titration with NaOH10 to 15 % w/v
Appearance (Form)Solution