Piperonyloyl chloride, 98%, Thermo Scientific Chemicals
Piperonyloyl chloride, 98%, Thermo Scientific Chemicals
Piperonyloyl chloride, 98%, Thermo Scientific Chemicals
Piperonyloyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperonyloyl chloride, 98%, Thermo Scientific Chemicals
CAS: 25054-53-9 | C8H5ClO3 | 184.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR370790100
Price (MYR)
2,625.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,625.00
Add to cart
Specifications
Chemical Name or MaterialPiperonyloyl chloride
CAS25054-53-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.