Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tributylphenyltin, Thermo Scientific Chemicals
CAS: 960-16-7 | C18H32Sn | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
Packaging:
Glass bottle
Catalog number ACR370230500
Price (MYR)
5,089.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,089.00
Add to cart
Specifications
Chemical Name or MaterialTributylphenyltin
CAS960-16-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Toxic if swallowed.
Harmful
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for se