3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
CAS: 107-86-8 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR370180250
Price (MYR)
1,371.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,371.00
Add to cart
Chemical Identifiers
CAS6106-24-7
IUPAC Namedisodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Molecular FormulaC4H8Na2O8
InChI KeyFGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILESO.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Optical Rotation26.3 ± 0.5? (C=1 in water)
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)12.22-18.84% (1.5-2.5 waters)
FormCrystals or powder or crystalline powder or fused solid