3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
CAS: 107-86-8 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR370180050
Price (MYR)
655.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
655.00
Add to cart
Chemical Identifiers
CAS97-74-5
IUPAC NameN,N-dimethyl[(dimethylcarbamothioyl)sulfanyl]carbothioamide
Molecular FormulaC6H12N2S3
InChI KeyREQPQFUJGGOFQL-UHFFFAOYSA-N
SMILESCN(C)C(=S)SC(=S)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (HPLC)≥96.0%
Melting Point (clear melt)104.0-114.0?C