2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
CAS: 31106-82-8 | C6H6BrN·HBr | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR370110250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77-99-6
IUPAC Name2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC6H14O3
InChI KeyZJCCRDAZUWHFQH-UHFFFAOYSA-N
SMILESCCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white
FormCrystals or powder or crystalline powder or flakes
Assay from Suppliers CofA≥97.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Free acid (titration)≤0.005% (as Formic acid)