Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminoguanidine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 1937-19-5 | CH6N4·HCl | 110.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR368911000
Price (MYR)
1,042.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,042.00
Add to cart
Specifications
Chemical Name or MaterialAminoguanidine hydrochloride
Name Note98%
CAS1937-19-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: