(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Triisopropylsilyl)acetylene, 97%, Thermo Scientific Chemicals
CAS: 89343-06-6 | C11H22Si | 182.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR368740050
Price (MYR)
727.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
727.00
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.94% (metals basis)
FormFoil, 0.025mm (0.001in) thick.
Assay from Supplier's CofA99.94% (metals basis)
Appearance (Color)Silvery
FormFoil, 0.025mm (0.001in) thick.