Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals
Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals
Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals
Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid, 20 wt.% solution in ethanol, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR368625000
Price (MYR)
1,251.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,251.00
Add to cart
Chemical Identifiers
CAS51429-74-4, 64-17-5
Specifications
Chemical Name or MaterialPhosphomolybdic acid
ColorGreen to Yellow
Name Note20 wt% solution in ethanol
CAS51429-74-4
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
For Manufacturing and Laboratory Use. Not for beverage use