Carvacrol, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Carvacrol, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR368341000100 g
Catalog number ACR368341000
Price (MYR)
884.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
884.00
Add to cart
Specifications
Chemical Name or MaterialCarvacrol
CAS499-75-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.