Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 mL
5 mL
25 mL
Catalog number ACR368121000
Price (MYR)
16,054.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
16,054.00
Add to cart
Chemical Identifiers
CAS93560-55-5
IUPAC Name2-iodo-3-methoxypyridine
Molecular FormulaC6H6INO
InChI KeyNJFRZBAZMPWJKQ-UHFFFAOYSA-N
SMILESCOC1=C(I)N=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)51.0-60.0?C
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%