Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerine trioleate, 98%, Thermo Scientific Chemicals
CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 mL
5 mL
100 mL
Catalog number ACR368120250
Price (MYR)
5,255.00
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
5,255.00
Add to cart
Specifications
Chemical Name or MaterialGlycerine trioleate
CAS122-32-7
ColorColorless to Yellow
Density0.9130g/mL
EINECS Number204-534-7