Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR368110050
Price (MYR)
4,255.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,255.00
Add to cart
Chemical Identifiers
CAS108-19-0
IUPAC Name1-(carbamoylamino)formamide
Molecular FormulaC2H5N3O2
InChI KeyOHJMTUPIZMNBFR-UHFFFAOYSA-N
SMILESNC(=O)NC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point184°C to 190°C
Appearance (Color)White
Assay>=96.0 % (ex N)
Loss on drying=<1 %