Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR368110010
Price (MYR)
1,120.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,120.00
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 2.0mm (0.08in) thick, annealed.
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silver