5'-Bromo-2'-hydroxyacetophenone, 98%
5'-Bromo-2'-hydroxyacetophenone, 98%
5'-Bromo-2'-hydroxyacetophenone, 98%
5'-Bromo-2'-hydroxyacetophenone, 98%
Thermo Scientific Chemicals
5'-Bromo-2'-hydroxyacetophenone, 98%
CAS: 1450-75-5 | C8H7BrO2 | 215.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR36721010010 gGlass bottle
Catalog number ACR367210100
Price (MYR)
612.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Specifications
Chemical Name or Material5′-Bromo-2′-hydroxyacetophenone
CAS1450-75-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0