Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene diacrylate, 95%, stabilized, Thermo Scientific Chemicals
CAS: 2274-11-5 | C8H10O4 | 170.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR36697025025 mLGlass bottle
Catalog number ACR366970250
Price (MYR)
3,110.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,110.00
EA
Add to cart
Chemical Identifiers
CAS6117-80-2
IUPAC Name(2Z)-but-2-ene-1,4-diol
Molecular FormulaC4H8O2
InChI KeyORTVZLZNOYNASJ-UPHRSURJSA-N
SMILESOC\C=C/CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.4750-1.4810 @ 20?C