Check Order Status
2,3-Dichlorophenylacetic acid, 97%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
2,3-Dichlorophenylacetic acid, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
ACR365550050
Price (MYR)
501.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Specifications
Chemical Name or Material
2, 3-Dichlorophenylacetic acid
CAS
10236-60-9
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0