Potassium tert-butoxide, pure, 20 wt.% solution in THF, Thermo Scientific Chemicals
Potassium tert-butoxide, pure, 20 wt.% solution in THF, Thermo Scientific Chemicals
Potassium tert-butoxide, pure, 20 wt.% solution in THF, Thermo Scientific Chemicals
Potassium tert-butoxide, pure, 20 wt.% solution in THF, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium tert-butoxide, pure, 20 wt.% solution in THF, Thermo Scientific Chemicals
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number ACR364990500
Price (MYR)
556.00
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Chemical Identifiers
CAS1314-61-0
IUPAC Name[(dioxotantalio)oxy]dioxotantalum
Molecular FormulaO5Ta2
InChI KeyPBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILESO=[Ta](=O)O[Ta](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Total Metal Impurities≤1%