n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR364970025
Price (MYR)
981.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
981.00
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.01 max. at 240nm, 0.02 max. at 230nm, 0.05 max. at 220nm, 0.1 max. at 210nm, 0.25 max. at 200nm
CAS142-82-5
ColorColorless
View more
For HPLC
RUO – Research Use Only