n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, for HPLC, (approx. 99% n-Heptane), Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR364970010
Price (MYR)
660.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
660.00
Add to cart
Chemical Identifiers
CAS6211-24-1
IUPAC Namebarium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
Molecular FormulaC24H20BaN2O6S2
InChI KeyIVCNVXFNTKXMCA-UHFFFAOYSA-L
SMILES[Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<1 % (105°C)
Sensitivity(as indicator) Passes test
For HPLC
RUO – Research Use Only