Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals
CAS: 5350-41-4 | C10H16BrN | 230.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR364860250
Price (MYR)
389.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
Add to cart
Chemical Identifiers
CAS34722-90-2
IUPAC Namesodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
Molecular FormulaC27H27Br2NaO5S
InChI KeyNMKFVGALBGZKGW-FKWCIMQXSA-M
SMILES[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to orange-brown
Infrared spectrumConforms
Visual transition intervalfrom pH 6.0 (yellow) to pH 7.6 (blue)
Appearance (Form)Powder
Clarity of solutionPasses test
Phase transfer catalyst
RUO – Research Use Only