Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 1L, Vapor Pressure: 20mbar at 20 deg.C, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR364425000
Price (MYR)
905.00
Add to cart
Quantity:
500 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
905.00
Add to cart
Chemical Identifiers
CAS118-46-7
IUPAC Name8-aminonaphthalen-2-ol
Molecular FormulaC10H9NO
InChI KeyKVHHMYZBFBSVDI-UHFFFAOYSA-N
SMILESNC1=C2C=C(O)C=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown
HPLC>=92 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals