Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
500 mL
2.5 L
Catalog number ACR364410010
Price (MYR)
697.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
697.00
Add to cart
Specifications
Chemical Name or MaterialToluene
Melting Point-95.0°C
Name Notewater <50 ppm
CAS108-88-3