Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR364350010
Price (MYR)
782.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
782.00
Add to cart
Chemical Identifiers
CAS64201-64-5
IUPAC Namequinolin-1-ium-1-olate
Molecular FormulaC9H7NO
InChI KeyGIIWGCBLYNDKBO-UHFFFAOYSA-N
SMILES[O-][N+]1=CC=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0% (dry wt. basis)
Water Content (Karl Fischer Titration)≤23.68%
Melting Point (clear melt)45.0-55.0?C