Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
500 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number ACR364330010
Price (MYR)
932.00
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
932.00
Add to cart
Chemical Identifiers
CAS102-76-1
IUPAC Name1,3-bis(acetyloxy)propan-2-yl acetate
Molecular FormulaC9H14O6
InChI KeyURAYPUMNDPQOKB-UHFFFAOYSA-N
SMILESCC(=O)OCC(COC(C)=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
Density(25°C) 1.154 to 1.158 g/ml