Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantity
ACR3643200252.5 L
Catalog number ACR364320025
Price (MYR)
1,668.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,668.00
EA
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
CAS67-66-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.