Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change view
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR364311000
Price (MYR)
953.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
953.00
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile
Melting Point-46.0°C
Name Notewater <50 ppm, extra dry over mol. sieve
CAS75-05-8
CAS Min %99.85