Zinc fluoride, 98%, Thermo Scientific Chemicals
Zinc fluoride, 98%, Thermo Scientific Chemicals
Zinc fluoride, 98%, Thermo Scientific Chemicals
Zinc fluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc fluoride, 98%, Thermo Scientific Chemicals
CAS: 7783-49-5 | F2Zn | 103.38 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR36391025025 gPlastic bottle
Catalog number ACR363910250
Price (MYR)
508.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Specifications
Boiling Point1500.0°C
Chemical Name or MaterialZinc fluoride
Melting Point872.0°C to 910.0°C
Name Note98%
CAS7783-49-5