Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Thermo Scientific Chemicals
Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ACR360112500
Price (MYR)
6,675.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
6,675.00
Add to cart
Specifications
PackagingGlass Bottle
Quantity250 g
MaterialSilica Gel
Unit Size
For column chromatography
RUO – Research Use Only