4-Thiouracil, 97%
4-Thiouracil, 97%
4-Thiouracil, 97%
Thermo Scientific Chemicals
4-Thiouracil, 97%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR359930010
Price (MYR)
1,120.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,120.00
Add to cart
Chemical Identifiers
CAS68528-80-3
IUPAC Namebis(2,5-dioxopyrrolidin-1-yl) octanedioate
Molecular FormulaC16H20N2O8
InChI KeyZWIBGKZDAWNIFC-UHFFFAOYSA-N
SMILESO=C(CCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
NMR (1H-NMR)Consistent with structure
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder