4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR359810050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7784-18-1
IUPAC Namealuminium(3+) trifluoride
Molecular FormulaAlF3
InChI KeyKLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)≤99.99% (metals basis)
FormCrystals or powder or crystalline powder
Total Metal Impurities≤0.01% (≤100ppm)
Elemental AnalysisAluminum, Al: Reported (Theory 32.13%)