Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR354800025
Price (MYR)
609.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
609.00
Add to cart
Chemical Identifiers
CAS54-64-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
Additional infoion mercuric: =< 0.7%
Loss on drying=<0.5 % (vacuum/P2O5)
Appearance (Form)Powder
View more
For peptide synthesis
RUO – Research Use Only