1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR354211000
Price (MYR)
2,223.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,223.00
Add to cart
Chemical Identifiers
CAS1871-57-4
IUPAC Name3-chloro-2-(chloromethyl)prop-1-ene
Molecular FormulaC4H6Cl2
InChI KeyXJFZOSUFGSANIF-UHFFFAOYSA-N
SMILESClCC(=C)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4750 to 1.4790 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=95 %
Appearance (Form)Liquid
Ionic liquid
RUO – Research Use Only