1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR354210050
Price (MYR)
718.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
718.00
Add to cart
Chemical Identifiers
CAS13939-06-5
IUPAC Namehexakis(methanidylidyneoxidanium) molybdenum
Molecular FormulaC6MoO6
InChI KeyKMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Elemental Analysis>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Ionic liquid
RUO – Research Use Only