1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Catalog number ACR354095000
Price (MYR)
4,436.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
4,436.00
Add to cart
Chemical Identifiers
CAS212713-08-1
IUPAC Namechlorobis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane
Molecular FormulaC30H46ClO2P
InChI KeyVOGHMZHRQUWLRE-UHFFFAOYSA-N
SMILESCOC1=C(C=C(C=C1C(C)(C)C)P(Cl)C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Proton NMRConforms to structure
Phosphorus-31 NMRConforms to structure
FormPowder
Assay by NMRPhosphorus P-31 NMR: ≥92.0%
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only