1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number ACR354091000
Price (MYR)
1,734.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,734.00
Add to cart
Specifications
Chemical Name or Material1-Butyl-3-methylimidazolium chloride
CAS79917-90-1
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only