1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Catalog number ACR354090250
Price (MYR)
769.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
769.00
Add to cart
Chemical Identifiers
CAS122049-54-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream or pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)4.4-13.2% (0.5-1.5 waters)
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only