1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number ACR354090050
Price (MYR)
620.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
620.00
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormRod, 5mm (0.2in) diameter.
Assay from Supplier's CofA≥99.999% (metals basis)
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only