4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals
4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals
4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals
4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals
CAS: 702-23-8 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR354050100
Price (MYR)
856.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
856.00
Add to cart
Chemical Identifiers
CAS4430-51-7
IUPAC Name1,4-diisothiocyanatobutane
Molecular FormulaC6H8N2S2
InChI KeyRRSISCMPUAGVJN-UHFFFAOYSA-N
SMILESS=C=NCCCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)30.0-36.0?C