Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 2855-13-2 | C10H22N2 | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR352942500
Price (MYR)
528.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
528.00
Add to cart
Chemical Identifiers
CAS1332-58-7
IUPAC Namedioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
Molecular FormulaAl2H4O9Si2
InChI KeyNLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILESO.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige
Loss on drying=<1.5 %
Particle size50 to 62 % (< 2 µm)
Appearance (Form)Powder
Loss on ignition10.8 % to 11.8 %