Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 2855-13-2 | C10H22N2 | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR352940010
Price (MYR)
958.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
958.00
Add to cart
Chemical Identifiers
CAS530-48-3
IUPAC Name(1-phenylethenyl)benzene
Molecular FormulaC14H12
InChI KeyZMYIIHDQURVDRB-UHFFFAOYSA-N
SMILESC=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6070 to 1.6090 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to golden
Infrared spectrumConforms
GC>=97.5 %