L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%
CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Catalog number ACR352685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS27532-96-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
TLC>=98.5 %